missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9'-Spirobi[9H-fluorene]-2-boronic Acid, TCI America™
$71.23 - $71.23
Specifications
| CAS | 236389-21-2 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C25H17BO2 |
| InChI Key | WDDLHUWVLROJLA-UHFFFAOYSA-N |
| PubChem CID | 22564369 |
Specifications
| 236389-21-2 | |
| C25H17BO2 | |
| 22564369 | |
| Crystalline Powder |
| White-Yellow | |
| WDDLHUWVLROJLA-UHFFFAOYSA-N | |
| 360.22 |