missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobi[9H-fluorene]-2-boronic Acid, TCI America™

Catalog No. S08311G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08311G Supplier TCI America Supplier No. S08311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 9,9′-Spirobi[9H-fluorene]-2-boronic Acid
CAS 236389-21-2
Color White-Yellow
Molecular Formula C25H17BO2
MDL Number MFCD13248568
Quantity 1 g
Synonym 9,9'-spirobi fluoren-2-ylboronic acid, 9,9'-spirobi 9h-fluorene-2-boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, 9,9'-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl, 9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, boronic acid, 9,9'-spirobi 9h-fluoren-2-yl, acmc-209g4k, 9,9'-spirobifluorene-2-boronic acid, 9,9'-spirobi fluorene-2-yl boronic acid
InChI Key WDDLHUWVLROJLA-UHFFFAOYSA-N
SMILES OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
IUPAC Name {9,9'-spirobi[fluoren]-2-yl}boronic acid
Molecular Weight (g/mol) 360.22
PubChem CID 22564369
Formula Weight 360.22
Physical Form Crystalline Powder
Show More Show Less