missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9'-Spirobi[9H-9-silafluorene] 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 232°C |
| Color | White |
| Boiling Point | 460°C |
| Quantity | 200 mg |
| Formula Weight | 332.48 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9,9′-Spirobi[9H-9-silafluorene] |
Safety and Handling
| TSCA | No |