accessibility menu, dialog, popup

UserName

CAS RN 159-68-2

Si

CAS RN 159-68-2

IUPAC Name: 8-sila-8,8'-spirobi[tricyclo[7.4.0.0²,⁷]tridecane]-1(13),1'(13'),2,2',4,4',6,6',9,9',11,11'-dodecaene
Synonyms: 8-sila-8,8'-spirobi[tricyclo[7.4.0.0²,⁷]tridecane]-1(13),1'(13'),2,2',4,4',6,6',9,9',11,11'-dodecaene
Molecular Weight (g/mol): 332.48
Molecular Formula: C24H16Si
InChi Key: FAMRBROKCVCCCX-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1[Si]21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results