missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Diphenylfluorene 98.0+%, TCI America™
$139.81 - $699.02
Chemical Identifiers
| CAS | 20302-14-1 |
|---|---|
| Molecular Formula | C25H18 |
| Molecular Weight (g/mol) | 318.419 |
| MDL Number | MFCD00093944 |
| InChI Key | BKQXUNGELBDWLS-UHFFFAOYSA-N |
| PubChem CID | 413569 |
| IUPAC Name | 9,9-diphenylfluorene |
| SMILES | C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=CC=C5 |
Chemical Identifiers
| 20302-14-1 | |
| 318.419 | |
| BKQXUNGELBDWLS-UHFFFAOYSA-N | |
| 9,9-diphenylfluorene |
| C25H18 | |
| MFCD00093944 | |
| 413569 | |
| C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=CC=C5 |
Specifications
| 20302-14-1 | |
| White | |
| MFCD00093944 | |
| BKQXUNGELBDWLS-UHFFFAOYSA-N | |
| 9,9-diphenylfluorene | |
| 413569 | |
| ≥98.0% (GC) | |
| 9,9-Diphenylfluorene |
| 226°C | |
| C25H18 | |
| 5 g | |
| C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=CC=C5 | |
| 318.419 | |
| 318.42 | |
| Crystalline Powder |
Safety and Handling
TSCA : No