missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole 98.0+%, TCI America™

Catalog No. D49035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49035G 5 g
D49031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49035G Supplier TCI America Supplier No. D49035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57102-62-2
Molecular Formula C36H24N2
Molecular Weight (g/mol) 484.602
InChI Key MHTPESFJWCJELK-UHFFFAOYSA-N
PubChem CID 23386665
IUPAC Name 9-phenyl-3-(9-phenylcarbazol-3-yl)carbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC5=C(C=C4)N(C6=CC=CC=C65)C7=CC=CC=C7)C8=CC=CC=C82

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 484.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′-Diphenyl-9H,9′H-3,3′-bicarbazole