accessibility menu, dialog, popup

UserName

NN

CAS RN 57102-62-2

IUPAC Name: 9,9'-diphenyl-9H,9'H-3,3'-bicarbazole
Synonyms: 9,9'-diphenyl-3,3'-bicarbazole
Molecular Weight (g/mol): 484.6
Molecular Formula: C36H24N2
InChi Key: MHTPESFJWCJELK-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC(=C2)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results