missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylfluorene 98.0+%, TCI America™

Catalog No. D32355G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32355G 5 g
D32351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32355G Supplier TCI America Supplier No. D32355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4569-45-3
Molecular Formula C15H14
Molecular Weight (g/mol) 194.28
MDL Number MFCD00114670,MFCD31699973
InChI Key ZHQNDEHZACHHTA-UHFFFAOYSA-N
PubChem CID 78325
IUPAC Name 9,9-dimethyl-9H-fluorene
SMILES CC1(C)C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 97°C
Color White
Boiling Point 287°C
Quantity 5 g
Formula Weight 194.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Dimethylfluorene