missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%

Catalog No. AAH5195003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5195003 1 g
AAH5195006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5195003 Supplier Thermo Scientific Chemicals Supplier No. H5195003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 569343-09-5 | C21H25BO2 | 320.239 g/mol

It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 569343-09-5
Molecular Formula C21H25BO2
Molecular Weight (g/mol) 320.239
MDL Number MFCD08704229
InChI Key DAZFRJAIIUPRQZ-UHFFFAOYSA-N
Synonym 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2-boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl, 2-9,9-dimethylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fl-be, amtb252, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9,9-dimethyl-9h-fluorene
PubChem CID 45358694
IUPAC Name 2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(C)C

Specifications

Quantity 1 g
Formula Weight 320.24
Percent Purity 95%
Chemical Name or Material 9,9-Dimethylfluorene-2-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only