missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%

Catalog No. p-7034284
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5195403 1 g
AAH5195406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5195403 Supplier Thermo Scientific Chemicals Supplier No. H5195403
Only null left

CAS: 325129-69-9 | C27H36B2O4 | 446.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 325129-69-9
Molecular Formula C27H36B2O4
Molecular Weight (g/mol) 446.201
MDL Number MFCD08704230
InChI Key RVFLMSKITNJVRB-UHFFFAOYSA-N
Synonym 2,2'-9,9-dimethyl-9h-fluorene-2,7-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2,7-diboronic acid bis pinacol ester, 2-9,9-dimethyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb256, 2,2-9,9-dimethyl-9h-fluorene-2,7-diyl, 9,9-dimethyl-9h-fluoren-2,7-diyl bisboronic acid pinacol ester, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9,9-dimethyl-9h-fluorene
PubChem CID 20640927
IUPAC Name 2-[9,9-dimethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(C)C)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Quantity 1 g
Formula Weight 446.2
Percent Purity 95%
Chemical Name or Material 9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only