missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylfluoren-2-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D39741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39741G Supplier TCI America Supplier No. D39741G

Specifications

Chemical Name or Material 9,9-Dimethylfluoren-2-boronic Acid (contains varying amounts of Anhydride)
CAS 333432-28-3
Color White-Yellow
Molecular Formula C15H15BO2
Quantity 1 g
MDL Number MFCD08704227
Synonym 9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid, 9,9-dimethylfluorene-2-boronic acid, 9,9-dimethyl-9h-fluorene-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronicacid
InChI Key DMDPAJOXRYGXCB-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
IUPAC Name (9,9-dimethylfluoren-2-yl)boronic acid
Molecular Weight (g/mol) 238.093
PubChem CID 22348454
Formula Weight 238.09
Physical Form Crystalline Powder
Show More Show Less