missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dihexylfluorene 98.0+%, TCI America™

Catalog No. D43275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43275G 5 g
D43271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43275G Supplier TCI America Supplier No. D43275G
Only null left

Chemical Identifiers

CAS 123863-97-8
Molecular Formula C25H34
Molecular Weight (g/mol) 334.55
MDL Number MFCD14584648
InChI Key LQQKFGSPUYTIRB-UHFFFAOYSA-N
PubChem CID 5128449
IUPAC Name 9,9-dihexyl-9H-fluorene
SMILES CCCCCCC1(CCCCCC)C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 32°C
Color White
Quantity 5 g
Formula Weight 334.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 9,9-Dihexylfluorene