missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Di-n-octylfluorene 97.0+%, TCI America™
Supplier: TCI America D32855G
Specifications
| 9,9-Di-n-octylfluorene | |
| C29H42 | |
| 5 g | |
| CCCCCCCCC1(CCCCCCCC)C2=CC=CC=C2C2=CC=CC=C12 | |
| 390.66 | |
| 390.66 | |
| Crystalline Lumps |
| 123863-99-0 | |
| MFCD08276358 | |
| RXACYPFGPNTUNV-UHFFFAOYSA-N | |
| 9,9-dioctyl-9H-fluorene | |
| 16213863 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 123863-99-0 | |
| 390.66 | |
| RXACYPFGPNTUNV-UHFFFAOYSA-N | |
| 9,9-dioctyl-9H-fluorene |
| C29H42 | |
| MFCD08276358 | |
| 16213863 | |
| CCCCCCCCC1(CCCCCCCC)C2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
TSCA : No