missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-methoxyphenyl)-9H-fluorene 98.0+%, TCI America™

Catalog No. B56175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56175G 5 g
B56171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56175G Supplier TCI America Supplier No. B56175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117766-40-2
Molecular Formula C27H22O2
Molecular Weight (g/mol) 378.471
InChI Key BYDIWLYAWBNCAQ-UHFFFAOYSA-N
PubChem CID 11783778
IUPAC Name 9,9-bis(4-methoxyphenyl)fluorene
SMILES COC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OC

Specifications

Melting Point 128°C
Color White
Quantity 5 g
Formula Weight 378.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-methoxyphenyl)-9H-fluorene