missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-hydroxyphenyl)fluorene 96.0+%, TCI America™

Catalog No. B171525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B171525G 25 g
B1715100G 100 g
B1715500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B171525G Supplier TCI America Supplier No. B171525G
Only null left

Chemical Identifiers

CAS 3236-71-3
Molecular Formula C25H18O2
Molecular Weight (g/mol) 350.42
MDL Number MFCD00191392
InChI Key YWFPGFJLYRKYJZ-UHFFFAOYSA-N
PubChem CID 76716
IUPAC Name 4-[9-(4-hydroxyphenyl)-9H-fluoren-9-yl]phenol
SMILES OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 227°C
Color White
Quantity 25 g
UN Number 3077
Synonym 4,4′C-(9-Fluorenylidene)diphenol
Formula Weight 350.42
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-hydroxyphenyl)fluorene
EINECSNumber (4)-1620
TSCA Yes