missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis(4-hydroxyphenyl)fluorene (purified by sublimation) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 226°C |
| Color | White |
| Quantity | 5 g |
| UN Number | 3077 |
| Synonym | 4,4′-(9-Fluorenylidene)diphenol |
| Formula Weight | 350.42 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9,9-Bis(4-hydroxyphenyl)fluorene (purified by sublimation) |
Safety and Handling
| EINECSNumber | (4)-1620 |
| TSCA | Yes |