missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis(4-aminophenyl)fluorene 98.0+%, TCI America™
$205.05 - $615.14
Chemical Identifiers
| CAS | 15499-84-0 |
|---|---|
| Molecular Formula | C25H20N2 |
| Molecular Weight (g/mol) | 348.449 |
| MDL Number | MFCD00039156 |
| InChI Key | KIFDSGGWDIVQGN-UHFFFAOYSA-N |
| PubChem CID | 631552 |
| IUPAC Name | 4-[9-(4-aminophenyl)fluoren-9-yl]aniline |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)N)C5=CC=C(C=C5)N |
Chemical Identifiers
| 15499-84-0 | |
| 348.449 | |
| KIFDSGGWDIVQGN-UHFFFAOYSA-N | |
| 4-[9-(4-aminophenyl)fluoren-9-yl]aniline |
| C25H20N2 | |
| MFCD00039156 | |
| 631552 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)N)C5=CC=C(C=C5)N |
Specifications
| 15499-84-0 | |
| White-Yellow | |
| MFCD00039156 | |
| 4,4′C-(9-Fluorenylidene)dianiline | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)N)C5=CC=C(C=C5)N | |
| 348.449 | |
| 348.45 | |
| Crystalline Powder |
| 236°C | |
| C25H20N2 | |
| 25 g | |
| KIFDSGGWDIVQGN-UHFFFAOYSA-N | |
| 4-[9-(4-aminophenyl)fluoren-9-yl]aniline | |
| 631552 | |
| ≥98.0% (T) | |
| 9,9-Bis(4-aminophenyl)fluorene |
Safety and Handling
EINECSNumber : (4)-1619
TSCA : No