missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-aminophenyl)fluorene 98.0+%, TCI America™

Catalog No. B154925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B154925G 25 g
B1549100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B154925G Supplier TCI America Supplier No. B154925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15499-84-0
Molecular Formula C25H20N2
Molecular Weight (g/mol) 348.449
MDL Number MFCD00039156
InChI Key KIFDSGGWDIVQGN-UHFFFAOYSA-N
PubChem CID 631552
IUPAC Name 4-[9-(4-aminophenyl)fluoren-9-yl]aniline
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)N)C5=CC=C(C=C5)N

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 25 g
Synonym 4,4′C-(9-Fluorenylidene)dianiline
Formula Weight 348.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-aminophenyl)fluorene
EINECSNumber (4)-1619
TSCA No