missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-amino-3-methylphenyl)fluorene 98.0+%, TCI America™

Catalog No. p-7119719
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26935G 5 g
B269325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26935G Supplier TCI America Supplier No. B26935G
Only null left

Chemical Identifiers

CAS 107934-60-1
Molecular Formula C27H24N2
Molecular Weight (g/mol) 376.503
InChI Key YRKVLGUIGNRYJX-UHFFFAOYSA-N
PubChem CID 21872150
IUPAC Name 4-[9-(4-amino-3-methylphenyl)fluoren-9-yl]-2-methylaniline
SMILES CC1=C(C=CC(=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC(=C(C=C5)N)C)N

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 5 g
Synonym 2,2′C-Dimethyl-4,4′C-(9-fluorenylidene)dianiline, 4,4′C-(9-Fluorenylidene)di-o-toluidine
Formula Weight 376.50
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-amino-3-methylphenyl)fluorene