missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene 98.0+%, TCI America™
$80.16 - $240.46
Chemical Identifiers
| CAS | 117344-32-8 |
|---|---|
| Molecular Formula | C29H26O4 |
| Molecular Weight (g/mol) | 438.52 |
| MDL Number | MFCD00216628 |
| InChI Key | NQXNYVAALXGLQT-UHFFFAOYSA-N |
| PubChem CID | 3764404 |
| IUPAC Name | 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]-9H-fluoren-9-yl}phenoxy)ethan-1-ol |
| SMILES | OCCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCCO)C=C1 |
Chemical Identifiers
| 117344-32-8 | |
| 438.52 | |
| NQXNYVAALXGLQT-UHFFFAOYSA-N | |
| 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]-9H-fluoren-9-yl}phenoxy)ethan-1-ol |
| C29H26O4 | |
| MFCD00216628 | |
| 3764404 | |
| OCCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCCO)C=C1 |
Specifications
| 117344-32-8 | |
| White-Yellow | |
| MFCD00216628 | |
| 4,4′C-(9-Fluorenylidene)bis(2-phenoxyethanol) | |
| OCCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCCO)C=C1 | |
| 438.52 | |
| 438.52 | |
| Crystalline Powder |
| 163°C | |
| C29H26O4 | |
| 5 g | |
| NQXNYVAALXGLQT-UHFFFAOYSA-N | |
| 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]-9H-fluoren-9-yl}phenoxy)ethan-1-ol | |
| 3764404 | |
| ≥98.0% (GC) | |
| 9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene |
Safety and Handling
EINECSNumber : (4)-1718
TSCA : No