missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene 98.0+%, TCI America™

Catalog No. F04475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04475G 5 g
F044725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04475G Supplier TCI America Supplier No. F04475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117344-32-8
Molecular Formula C29H26O4
Molecular Weight (g/mol) 438.52
MDL Number MFCD00216628
InChI Key NQXNYVAALXGLQT-UHFFFAOYSA-N
PubChem CID 3764404
IUPAC Name 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]-9H-fluoren-9-yl}phenoxy)ethan-1-ol
SMILES OCCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCCO)C=C1

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 5 g
Synonym 4,4′C-(9-Fluorenylidene)bis(2-phenoxyethanol)
Formula Weight 438.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene
EINECSNumber (4)-1718
TSCA No