missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(3,4-dihydroxyphenyl)fluorene 98.0+%, TCI America™

Catalog No. p-7123678
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B46651G 1 g
B46655G 5 g
2 options

Catalog No. B46651G

Supplier: TCI America B46651G

Only null left

Chemical Identifiers

CAS 351521-78-3
Molecular Formula C25H18O4
Molecular Weight (g/mol) 382.42
MDL Number MFCD31618097
InChI Key RAIOQXXWEGZKAI-UHFFFAOYSA-N
Synonym 4,4′-(9H-Fluorene-9,9-diyl)bis(benzene-1,2-diol)
PubChem CID 24778256
IUPAC Name 4-[9-(3,4-dihydroxyphenyl)-9H-fluoren-9-yl]benzene-1,2-diol
SMILES OC1=C(O)C=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(O)=C(O)C=C1

Specifications

Melting Point 183°C
Color White
Quantity 1 g
Formula Weight 382.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(3,4-dihydroxyphenyl)fluorene