missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene 98.0+%, TCI America™

Catalog No. p-7123821
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4892200MG 200 mg
B48921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4892200MG Supplier TCI America Supplier No. B4892200MG

Chemical Identifiers

CAS 357219-41-1
Molecular Formula C41H64B2O4
Molecular Weight (g/mol) 642.579
MDL Number MFCD16621099
InChI Key SMQUVDNJQVDOLI-UHFFFAOYSA-N
Synonym 9,9-Bis(2-ethylhexyl)fluorene-2,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 12104084
IUPAC Name 2-[9,9-bis(2-ethylhexyl)-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(CC(CC)CCCC)CC(CC)CCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 200 mg
Formula Weight 642.58
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene