missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Bromophenyl)-9-phenylfluorene 98.0+%, TCI America™

Catalog No. B52211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52211G 1 g
B52215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52211G Supplier TCI America Supplier No. B52211G
Only null left

Chemical Identifiers

CAS 937082-81-0
Molecular Formula C25H17Br
Molecular Weight (g/mol) 397.315
MDL Number MFCD28975087
InChI Key OOKRYIPMHLUQHU-UHFFFAOYSA-N
PubChem CID 57746345
IUPAC Name 9-(4-bromophenyl)-9-phenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)Br

Specifications

Melting Point 153°C
Color White
Quantity 1 g
Formula Weight 397.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Bromophenyl)-9-phenylfluorene