missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole 98.0+%, TCI America™

Catalog No. B53135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53135G 5 g
B53131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53135G Supplier TCI America Supplier No. B53135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 601454-33-5
Molecular Formula C26H28BrN
Molecular Weight (g/mol) 434.421
MDL Number MFCD11617971
InChI Key LAHSPJHDQUQBTB-UHFFFAOYSA-N
PubChem CID 57604191
IUPAC Name 9-(4-bromophenyl)-3,6-ditert-butylcarbazole
SMILES CC(C)(C)C1=CC2=C(C=C1)N(C3=C2C=C(C=C3)C(C)(C)C)C4=CC=C(C=C4)Br

Specifications

Melting Point 156°C
Color White
Quantity 5 g
Formula Weight 434.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole