accessibility menu, dialog, popup

UserName

CH3CH3H3CNH3CH3CCH3Br

CAS RN 601454-33-5

IUPAC Name: 9-(4-bromophenyl)-3,6-di-tert-butyl-9H-carbazole
Synonyms: 9-(4-bromophenyl)-3,6-di-tert-butylcarbazole
Molecular Weight (g/mol): 434.42
Molecular Formula: C26H28BrN
InChi Key: LAHSPJHDQUQBTB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C(C)(C)C)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results