missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Biphenylyl)-10-bromoanthracene 98.0+%, TCI America™

Catalog No. B45695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45695G 5 g
B45691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45695G Supplier TCI America Supplier No. B45695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 400607-05-8
Molecular Formula C26H17Br
Molecular Weight (g/mol) 409.33
MDL Number MFCD19440858
InChI Key VCJIOUBBOCVHPE-UHFFFAOYSA-N
PubChem CID 21076372
IUPAC Name 9-{[1,1'-biphenyl]-4-yl}-10-bromoanthracene
SMILES BrC1=C2C=CC=CC2=C(C2=CC=C(C=C2)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 253°C
Color Yellow
Quantity 5 g
Formula Weight 409.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Biphenylyl)-10-bromoanthracene