accessibility menu, dialog, popup

UserName

CAS RN 400607-05-8

IUPAC Name: 9-{[1,1'-biphenyl]-4-yl}-10-bromoanthracene
Synonyms: 9-{[1,1'-biphenyl]-4-yl}-10-bromoanthracene
Molecular Weight (g/mol): 409.33
Molecular Formula: C26H17Br
InChi Key: VCJIOUBBOCVHPE-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=C(C=C2)C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results