missing translation for 'onlineSavingsMsg'
Learn More

9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole 98.0+%, TCI America™

$262.83 - $784.76

Chemical Identifiers

CAS 1338446-77-7
Molecular Formula C30H19NO
Molecular Weight (g/mol) 409.488
InChI Key YEWVLWWLYHXZLZ-UHFFFAOYSA-N
PubChem CID 58556666
IUPAC Name 9-(3-dibenzofuran-2-ylphenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=CC(=C4)C5=CC6=C(C=C5)OC7=CC=CC=C76
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D4919200MG
View Documents
TCI America
D4919200MG
200 mg
Each for $262.83
Only null left
 
D49191G
View Documents
TCI America
D49191G
1 g
Each for $784.76
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1338446-77-7
409.488
58556666
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=CC(=C4)C5=CC6=C(C=C5)OC7=CC=CC=C76
C30H19NO
YEWVLWWLYHXZLZ-UHFFFAOYSA-N
9-(3-dibenzofuran-2-ylphenyl)carbazole

Specifications

1338446-77-7
C30H19NO
YEWVLWWLYHXZLZ-UHFFFAOYSA-N
9-(3-dibenzofuran-2-ylphenyl)carbazole
58556666
≥98.0% (HPLC,N)
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole
White-Yellow
200 mg
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=CC(=C4)C5=CC6=C(C=C5)OC7=CC=CC=C76
409.488
409.49
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents