missing translation for 'onlineSavingsMsg'
Learn More

9-(3-Bromophenyl)-9-phenyl-9H-fluorene 98.0+%, TCI America™

Catalog No. B56165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56165G 5 g
B56161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56165G Supplier TCI America Supplier No. B56165G

Chemical Identifiers

CAS 1257251-75-4
Molecular Formula C25H17Br
Molecular Weight (g/mol) 397.315
MDL Number MFCD30187277
InChI Key LWBYEDXZPBXTFL-UHFFFAOYSA-N
PubChem CID 58233549
IUPAC Name 9-(3-bromophenyl)-9-phenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC(=CC=C5)Br

Specifications

Melting Point 133°C
Color White
Quantity 5 g
Formula Weight 397.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(3-Bromophenyl)-9-phenyl-9H-fluorene