missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(2'-Bromo-4-biphenylyl)carbazole 98.0+%, TCI America™
Supplier: TCI America B5312200MG
Specifications
| 9-(2′-Bromo-4-biphenylyl)carbazole | |
| 106°C | |
| 200 mg | |
| 398.30 | |
| Crystalline Powder |
| 1215228-57-1 | |
| C24H16BrN | |
| 9-(2′-Bromo-[1,1′-biphenyl]-4-yl)-9H-carbazole | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1215228-57-1 | |
| C24H16BrN | |
| 9-(2′-Bromo-[1,1′-biphenyl]-4-yl)-9H-carbazole |
Safety and Handling
TSCA : No