missing translation for 'onlineSavingsMsg'
Learn More

9-(2'-Bromo-4-biphenylyl)carbazole 98.0+%, TCI America™

Catalog No. p-7124037
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5312200MG 200 mg
B53121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5312200MG Supplier TCI America Supplier No. B5312200MG
Only null left

Chemical Identifiers

CAS 1215228-57-1
Molecular Formula C24H16BrN
Synonym 9-(2′-Bromo-[1,1′-biphenyl]-4-yl)-9H-carbazole

Specifications

Melting Point 106°C
Quantity 200 mg
Formula Weight 398.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(2′-Bromo-4-biphenylyl)carbazole