missing translation for 'onlineSavingsMsg'
Learn More

9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole 98.0+%, TCI America™

Catalog No. B19675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19675G 5 g
B19671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19675G Supplier TCI America Supplier No. B19675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124337-34-4
Molecular Formula C19H14N4
Molecular Weight (g/mol) 298.349
MDL Number MFCD00216631
InChI Key JWLQIMHJRKCIFA-UHFFFAOYSA-N
PubChem CID 4519736
IUPAC Name 9-(benzotriazol-1-ylmethyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2CN4C5=CC=CC=C5N=N4

Specifications

Quantity 5 g
Formula Weight 298.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole