accessibility menu, dialog, popup

UserName

CAS RN 124337-34-4

NNNN

CAS RN 124337-34-4

IUPAC Name: 9-[(1H-1,2,3-benzotriazol-1-yl)methyl]-9H-carbazole
Synonyms: 9-(1,2,3-benzotriazol-1-ylmethyl)carbazole
Molecular Weight (g/mol): 298.35
Molecular Formula: C19H14N4
InChi Key: JWLQIMHJRKCIFA-UHFFFAOYSA-N
SMILES: C(N1N=NC2=CC=CC=C12)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results