missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9(10H)-Acridone 98.0+%, TCI America™
Supplier: TCI America A01335G
Specifications
| 9(10H)-Acridone | |
| 354°C | |
| C13H9NO | |
| 5 g | |
| FZEYVTFCMJSGMP-UHFFFAOYSA-N | |
| 10H-acridin-9-one | |
| 2015 | |
| 195.22 | |
| Crystalline Powder |
| 578-95-0 | |
| Yellow | |
| MFCD00005019 | |
| acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine | |
| C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2 | |
| 195.221 | |
| CHEBI:50756 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 578-95-0 | |
| 195.221 | |
| FZEYVTFCMJSGMP-UHFFFAOYSA-N | |
| 2015 | |
| 10H-acridin-9-one |
| C13H9NO | |
| MFCD00005019 | |
| acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine | |
| CHEBI:50756 | |
| C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2 |
Safety and Handling
RTECSNumber : AR6976000
TSCA : Yes