missing translation for 'onlineSavingsMsg'
Learn More

9(10H)-Acridone 98.0+%, TCI America™

Catalog No. p-7116732
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
A01335G 5 g
A013325G 25 g
2 options

Catalog No. A01335G

Supplier: TCI America A01335G

Only null left

Chemical Identifiers

CAS 578-95-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00005019
InChI Key FZEYVTFCMJSGMP-UHFFFAOYSA-N
Synonym acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
PubChem CID 2015
ChEBI CHEBI:50756
IUPAC Name 10H-acridin-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2

Specifications

Melting Point 354°C
Color Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9(10H)-Acridone
RTECSNumber AR6976000
TSCA Yes