missing translation for 'onlineSavingsMsg'
Learn More

9,10-Phenanthrenequinone 99.0+%, TCI America™

Catalog No. p-7134413
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00805G 5 g
P008025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00805G Supplier TCI America Supplier No. P00805G
Only null left

Chemical Identifiers

CAS 84-11-7
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.216
MDL Number MFCD00001163
InChI Key YYVYAPXYZVYDHN-UHFFFAOYSA-N
Synonym 9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
PubChem CID 6763
ChEBI CHEBI:37454
IUPAC Name phenanthrene-9,10-dione
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O

Specifications

Melting Point 210°C
Quantity 5 g
UN Number 3077
Formula Weight 208.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Phenanthrenequinone
RTECSNumber SF7875000
TSCA Yes