missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Diphenylanthracene, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 1499-10-1 |
| Physical Form | Neat |
| Linear Formula | C26H18 |
| MDL Number | MFCD00001253 |
| Melting Point | 245°C to 248°C |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 330.42 |
| Grade | Analytical Standard |