missing translation for 'onlineSavingsMsg'
Learn More

9,10-Diphenylanthracene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7128386
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4647200MG 200 mg
D46471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4647200MG Supplier TCI America Supplier No. D4647200MG
Only null left

Chemical Identifiers

CAS 597553-98-5
Molecular Formula C26H19BO2
Molecular Weight (g/mol) 374.246
MDL Number MFCD16621110
InChI Key MVUDLJXJTYSUGF-UHFFFAOYSA-N
PubChem CID 23160215
IUPAC Name (9,10-diphenylanthracen-2-yl)boronic acid
SMILES B(C1=CC2=C(C3=CC=CC=C3C(=C2C=C1)C4=CC=CC=C4)C5=CC=CC=C5)(O)O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 374.25
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Diphenylanthracene-2-boronic Acid (contains varying amounts of Anhydride)