accessibility menu, dialog, popup

UserName

CAS RN 597553-98-5

IUPAC Name: (9,10-diphenylanthracen-2-yl)boronic acid
Synonyms: 9,10-diphenylanthracen-2-ylboronic acid
Molecular Weight (g/mol): 374.25
Molecular Formula: C26H19BO2
InChi Key: MVUDLJXJTYSUGF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C(=C1)C(C1=CC=CC=C1)=C1C=CC=CC1=C2C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results