missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dimethylanthracene 98.0+%, TCI America™

Catalog No. D02521G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02521G 1 g
D025210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02521G Supplier TCI America Supplier No. D02521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

Specifications

Melting Point 182°C
Color Yellow
Quantity 1 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dimethylanthracene
EINECSNumber (4)-0640
RTECSNumber CA9685000
TSCA No