missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dimethylanthracene, 97%

Catalog No. AAB2245806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2245806 5 g
AAB2245803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2245806 Supplier Thermo Scientific Chemicals Supplier No. B2245806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 781-43-1 | C16H14 | 206.29 g/mol

9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

Specifications

Melting Point 182°C to 184°C
Quantity 5 g
Beilstein 1909028
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 9,10-Dimethylanthracene
Hazard Category Warning
Hazard Statement GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-308-4
RTECSNumber CA9685000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only