missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dicyanoanthracene 98.0+%, TCI America™

Catalog No. p-7126856
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D16561G 1 g
D16565G 5 g
2 options

Catalog No. D16561G

Supplier: TCI America D16561G

Only null left

Chemical Identifiers

CAS 1217-45-4
Molecular Formula C16H8N2
Molecular Weight (g/mol) 228.25
MDL Number MFCD00041733
InChI Key BIOPPFDHKHWJIA-UHFFFAOYSA-N
Synonym 9,10-dicyanoanthracene, 9,10-anthracenedicarbonitrile, 9,10-dicyanoanthracen, anthracene-9,10-dinitrile, acmc-209af1
PubChem CID 71035
ChEBI CHEBI:51302
IUPAC Name anthracene-9,10-dicarbonitrile
SMILES N#CC1=C2C=CC=CC2=C(C#N)C2=CC=CC=C12

Specifications

Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 228.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dicyanoanthracene