missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Dicyanoanthracene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 228.25 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9,10-Dicyanoanthracene |
Safety and Handling
| TSCA | Yes |