missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dichloroanthracene 96.0+%, TCI America™

Catalog No. p-7126147
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03271G 1 g
D032710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03271G Supplier TCI America Supplier No. D03271G
Only null left

Chemical Identifiers

CAS 605-48-1
Molecular Formula C14H8Cl2
Molecular Weight (g/mol) 247.118
MDL Number MFCD00001246
InChI Key FKDIWXZNKAZCBY-UHFFFAOYSA-N
Synonym anthracene, 9,10-dichloro, acmc-209mkk, anthracene,10-dichloro, 9,10-dichloro-anthracene, anthracene,9,10-dichloro, 9.10-dichloroanthracene
PubChem CID 11800
IUPAC Name 9,10-dichloroanthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Cl)Cl

Specifications

Melting Point 210°C
Quantity 1 g
Formula Weight 247.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dichloroanthracene