missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dibromo-2-methylanthracene 96.0+%, TCI America™

Catalog No. D4977200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4977200MG 200 mg
D49771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4977200MG Supplier TCI America Supplier No. D4977200MG
Only null left

Chemical Identifiers

CAS 177839-45-1
Molecular Formula C15H10Br2
Molecular Weight (g/mol) 350.05
MDL Number MFCD00266492
InChI Key PNDNWMMKRSTYTC-UHFFFAOYSA-N
PubChem CID 1713096
IUPAC Name 9,10-dibromo-2-methylanthracene
SMILES CC1=CC=C2C(Br)=C3C=CC=CC3=C(Br)C2=C1

Specifications

Melting Point 143°C
Color Yellow
Quantity 200 mg
Formula Weight 350.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dibromo-2-methylanthracene