missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Di(2-naphthyl)anthracene 98.0+%, TCI America™
$292.65 - $292.65
Chemical Identifiers
| CAS | 122648-99-1 |
|---|---|
| Molecular Formula | C34H22 |
| Molecular Weight (g/mol) | 430.55 |
| MDL Number | MFCD00028944 |
| InChI Key | VIZUPBYFLORCRA-UHFFFAOYSA-N |
| PubChem CID | 21881242 |
| IUPAC Name | 9,10-dinaphthalen-2-ylanthracene |
| SMILES | C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=CC=CC=C7C=C6 |
Chemical Identifiers
| 122648-99-1 | |
| 430.55 | |
| VIZUPBYFLORCRA-UHFFFAOYSA-N | |
| 9,10-dinaphthalen-2-ylanthracene |
| C34H22 | |
| MFCD00028944 | |
| 21881242 | |
| C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=CC=CC=C7C=C6 |
Specifications
| 122648-99-1 | |
| Yellow | |
| MFCD00028944 | |
| VIZUPBYFLORCRA-UHFFFAOYSA-N | |
| 9,10-dinaphthalen-2-ylanthracene | |
| 21881242 | |
| ≥98.0% (HPLC) | |
| 9,10-Di(2-naphthyl)anthracene |
| 357°C | |
| C34H22 | |
| 1 g | |
| C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=CC=CC=C7C=C6 | |
| 430.55 | |
| 430.55 | |
| Crystalline Powder |
Safety and Handling
TSCA : No