missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(phenylethynyl)anthracene

Catalog No. p-7058799
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4331303 1 g
AA4331306 5 g
AA4331314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4331303 Supplier Thermo Scientific Chemicals Supplier No. 04331303
Only null left

As a photochromic material exhibiting chemiluminescence | CAS: 10075-85-1 | C30H18 | 378.474 g/mol

9,10-Bis(phenylethynyl)anthracene is used as a chemiluminescent fluorophore with high quantum efficiency. It is used in lightsticks as a fluorophor producing ghostly green light. It is also used as a dopant for organic semiconductors in OLEDs. It is used as a reagent for chemiluminescence research. 9,10-Bis(phenylethynyl)anthracene is known to exert fluorescence properties due to its polycyclic aromatic hydrocarbon system and it is commonly used as an scintillator additive. 9,10-Bis(phenylethynyl)anthracene was also studied extensively as a fluorescence emitters for peroxyoxalate chemiluminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10075-85-1
Molecular Formula C30H18
Molecular Weight (g/mol) 378.474
MDL Number MFCD00012050
InChI Key ZHBOFZNNPZNWGB-UHFFFAOYSA-N
Synonym 9,10-bis phenylethynyl anthracene, 9,10-bis 2-phenylethynyl anthracene, anthracene, 9,10-bis phenylethynyl, bpea, anthracene, 9,10-bis 2-phenylethynyl, bis phcc-anthracene, unii-fc8jdb70dq, 9,10-bis phenylvinyl anthracene, fc8jdb70dq, 9,10-bis-phenylethynyl anthracene
PubChem CID 82338
ChEBI CHEBI:51675
IUPAC Name 9,10-bis(2-phenylethynyl)anthracene
SMILES C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5

Specifications

Melting Point 254°C to 255°C
Quantity 1 g
Beilstein 1891432
Sensitivity Light sensitive
Solubility Information It is insoluble in water but is soluble in most organic solvents such as carbon disulfide,alcohols,benzene,chloroform,and hydronaphthalenes.
Formula Weight 378.47
Physical Form Crystalline
Chemical Name or Material 9,10-Bis (phenylethynyl)anthracene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-210-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only