missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(phenylethynyl)anthracene 98.0+%, TCI America™

Catalog No. P11371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11371G 1 g
P11375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11371G Supplier TCI America Supplier No. P11371G

Chemical Identifiers

CAS 10075-85-1
Molecular Formula C30H18
Molecular Weight (g/mol) 378.474
MDL Number MFCD00012050
InChI Key ZHBOFZNNPZNWGB-UHFFFAOYSA-N
Synonym 9,10-bis phenylethynyl anthracene, 9,10-bis 2-phenylethynyl anthracene, anthracene, 9,10-bis phenylethynyl, bpea, anthracene, 9,10-bis 2-phenylethynyl, bis phcc-anthracene, unii-fc8jdb70dq, 9,10-bis phenylvinyl anthracene, fc8jdb70dq, 9,10-bis-phenylethynyl anthracene
PubChem CID 82338
ChEBI CHEBI:51675
IUPAC Name 9,10-bis(2-phenylethynyl)anthracene
SMILES C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5

Specifications

Melting Point 253°C
Quantity 1 g
Formula Weight 378.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(phenylethynyl)anthracene