missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(chloromethyl)anthracene 97.0+%, TCI America™

Catalog No. B13275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13275G 5 g
B132725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13275G Supplier TCI America Supplier No. B13275G
Only null left

Chemical Identifiers

CAS 10387-13-0
Molecular Formula C16H12Cl2
Molecular Weight (g/mol) 275.172
MDL Number MFCD00045388
InChI Key UOSROERWQJTVNU-UHFFFAOYSA-N
PubChem CID 25219
IUPAC Name 9,10-bis(chloromethyl)anthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2CCl)CCl

Specifications

Quantity 5 g
UN Number 3261
Formula Weight 275.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(chloromethyl)anthracene
RTECSNumber CA9365000
TSCA Yes